ENA429360143 Enamine

sodium 2-(isoquinolin-1-yl)acetate

  • MDL number MFCD28118836
  • Cas Number : 1798728-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189452
Availability details for
Quantity
  • Availability
  • Add to Cart