ENA429360146 Enamine

methyl 2-chloro-2-(thiophen-3-yl)acetate

  • MDL number MFCD16694217
  • Cas Number : 1249784-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189455
Availability details for
Quantity
  • Availability
  • Add to Cart