ENA429360324 Enamine

6-(2,6-dimethylmorpholin-4-yl)pyridin-3-amine dihydrochloride

  • MDL number MFCD28125052
  • Cas Number : 1803586-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189894
Availability details for
Quantity
  • Availability
  • Add to Cart