ENA429360325 Enamine

3-(4-iodo-3,5-dimethyl-1H-pyrazol-1-yl)-1lambda6-thiolane-1,1-dione hydrochloride

  • MDL number MFCD28125053
  • Cas Number : 1788823-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189899
Availability details for
Quantity
  • Availability
  • Add to Cart