ENA429360333 Enamine

2-[(2-aminoethyl)(cyclopropyl)amino]ethan-1-ol dihydrochloride

  • MDL number MFCD28125064
  • Cas Number : 1803600-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190138
Availability details for
Quantity
  • Availability
  • Add to Cart