ENA429360347 Enamine

4-(2-aminoethyl)-3-methylpiperazin-2-one trihydrochloride

  • MDL number MFCD28125082
  • Cas Number : 1803571-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190297
Availability details for
Quantity
  • Availability
  • Add to Cart