ENA429360352 Enamine

dimethyl({2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]ethyl})amine hydrochloride

  • MDL number MFCD28125089

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190419
Availability details for
Quantity
  • Availability
  • Add to Cart