ENA429360362 Enamine

2-(3-carboxyprop-2-enamido)-5-nitrobenzoic acid

  • Cas Number : 1807940-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190519
Availability details for
Quantity
  • Availability
  • Add to Cart