ENA429360364 Enamine

1-(1H-pyrazol-3-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD28125102
  • Cas Number : 1803571-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190542
Availability details for
Quantity
  • Availability
  • Add to Cart