ENA429360365 Enamine

2-amino-3-(benzylamino)propanoic acid dihydrochloride

  • MDL number MFCD28125103
  • Cas Number : 1786218-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190558
Availability details for
Quantity
  • Availability
  • Add to Cart