ENA429360372 Enamine

9-chloro-1H,2H,3H,4H,5H,6H-azepino[4,3-b]indole hydrochloride

  • MDL number MFCD28125109
  • Cas Number : 1803609-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190674
Availability details for
Quantity
  • Availability
  • Add to Cart