ENA429360375 Enamine

4-[(3-methylphenyl)methyl]piperidin-4-ol hydrochloride

  • MDL number MFCD28125112
  • Cas Number : 1795502-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-190701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190701
Availability details for
Quantity
  • Availability
  • Add to Cart