ENA429360386 Enamine

4-methyl-2-(1H-1,2,4-triazol-1-yl)cyclohexan-1-one

  • MDL number MFCD28125123
  • Cas Number : 1788567-07-6
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-190854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190854
Availability details for
Quantity
  • Availability
  • Add to Cart