ENA429360401 Enamine

1-(3-aminopropyl)-1,2-dihydropyrimidin-2-one dihydrochloride

  • MDL number MFCD28125138
  • Cas Number : 1803607-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191114
Availability details for
Quantity
  • Availability
  • Add to Cart