ENA429360409 Enamine

[(2,2-dimethyl-1,3-dioxolan-4-yl)methyl][(4-methoxyphenyl)methyl]amine hydrochloride

  • MDL number MFCD28125148
  • Cas Number : 1803600-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191223
Availability details for
Quantity
  • Availability
  • Add to Cart