ENA429360419 Enamine

N-(2-aminoethyl)cyclobutanecarboxamide hydrochloride

  • MDL number MFCD28125163
  • Cas Number : 1803599-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191323
Availability details for
Quantity
  • Availability
  • Add to Cart