ENA429360420 Enamine

3-(5-amino-2-fluorophenyl)-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD28125164
  • Cas Number : 1788989-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191331
Availability details for
Quantity
  • Availability
  • Add to Cart