ENA429360422 Enamine

3-(2-aminoethyl)-2-methyl-3,4-dihydroquinazolin-4-one dihydrochloride

  • MDL number MFCD28125166
  • Cas Number : 1803583-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191335
Availability details for
Quantity
  • Availability
  • Add to Cart