ENA429360423 Enamine

methyl 3-[(pyrimidin-2-yl)methanesulfonyl]propanoate

  • MDL number MFCD28125167
  • Cas Number : 1787856-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191359
Availability details for
Quantity
  • Availability
  • Add to Cart