ENA429360436 Enamine

ethyl 5-acetyl-2-amino-1,3-thiazole-4-carboxylate hydrochloride

  • MDL number MFCD28125182
  • Cas Number : 1798747-14-4
  • Molecular Weight : 250.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-191499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191499
Availability details for
Quantity
  • Availability
  • Add to Cart