ENA429360446 Enamine

3,5-dichloro-4-sulfanylphenol

  • MDL number MFCD28125193
  • Cas Number : 1803611-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191705
Availability details for
Quantity
  • Availability
  • Add to Cart