ENA429360456 Enamine

4-chloro-2-(methoxymethyl)aniline hydrochloride

  • MDL number MFCD28125205
  • Cas Number : 1803610-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191933
Availability details for
Quantity
  • Availability
  • Add to Cart