ENA429360457 Enamine

N,N-dimethyl-2,3-dihydro-1H-isoindole-5-sulfonamide hydrochloride

  • MDL number MFCD28125206
  • Cas Number : 1798733-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191936
Availability details for
Quantity
  • Availability
  • Add to Cart