ENA429360459 Enamine

3-[(pentan-2-yl)amino]benzonitrile hydrochloride

  • MDL number MFCD28125208
  • Cas Number : 1803566-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191998
Availability details for
Quantity
  • Availability
  • Add to Cart