ENA429360463 Enamine

imidazo[1,2-a]pyridin-7-amine hydrochloride

  • MDL number MFCD28125212
  • Cas Number : 1795435-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192009

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192009
Availability details for
Quantity
  • Availability
  • Add to Cart