ENA429360472 Enamine

1-(3-fluoro-5-methylphenyl)-1H-1,2,3-triazol-4-amine hydrochloride

  • MDL number MFCD28125221
  • Cas Number : 1788627-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192136
Availability details for
Quantity
  • Availability
  • Add to Cart