ENA429360473 Enamine

4-(4-methylphenyl)piperidine-4-carboxylic acid hydrochloride

  • MDL number MFCD28125222
  • Cas Number : 1803596-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192162
Availability details for
Quantity
  • Availability
  • Add to Cart