ENA429360481 Enamine

ethyl 2-(3-fluoropyridin-2-yl)propanoate

  • MDL number MFCD28125232
  • Cas Number : 1803591-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192268
Availability details for
Quantity
  • Availability
  • Add to Cart