ENA429360483 Enamine

methyl 2-{1H-pyrrolo[2,3-c]pyridin-3-yl}acetate

  • MDL number MFCD28125234
  • Cas Number : 1803571-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192273
Availability details for
Quantity
  • Availability
  • Add to Cart