ENA429360484 Enamine

3,3-diethyl 4-hydroxy-1-oxo-3,4-dihydro-1H-2-benzopyran-3,3-dicarboxylate

  • MDL number MFCD28125235
  • Cas Number : 95168-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192274
Availability details for
Quantity
  • Availability
  • Add to Cart