ENA429360485 Enamine

2-(4-{[(2-chloroethoxy)carbonyl]amino}phenyl)acetic acid

  • MDL number MFCD28125236
  • Cas Number : 437982-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192324
Availability details for
Quantity
  • Availability
  • Add to Cart