ENA429360489 Enamine

methyl 2-amino-3-(prop-2-en-1-yloxy)propanoate hydrochloride

  • MDL number MFCD28125240
  • Cas Number : 2170123-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192382
Availability details for
Quantity
  • Availability
  • Add to Cart