ENA429360491 Enamine

2-methoxy-4-(methylsulfanyl)-1-nitrobenzene

  • MDL number MFCD22489310
  • Cas Number : 860734-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189904
Availability details for
Quantity
  • Availability
  • Add to Cart