ENA429360499 Enamine

2-methyl-2H-indazole-3-carbonitrile

  • MDL number MFCD09753443
  • Cas Number : 31748-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189487

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189487
Availability details for
Quantity
  • Availability
  • Add to Cart