ENA429360500 Enamine

2-methyl-1,9-dioxa-4-azaspiro[5.5]undecane

  • MDL number MFCD21127021
  • Cas Number : 1481966-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189957
Availability details for
Quantity
  • Availability
  • Add to Cart