ENA429360502 Enamine

6-bromo-4-chloropyridine-3-carbaldehyde

  • MDL number MFCD13188856
  • Cas Number : 1060811-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189959
Availability details for
Quantity
  • Availability
  • Add to Cart