ENA429360503 Enamine

N,N,5-trimethyl-1H-pyrazole-4-sulfonamide

  • MDL number MFCD12804951

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189963
Availability details for
Quantity
  • Availability
  • Add to Cart