ENA429360505 Enamine

1-[2-fluoro-4-(furan-2-yl)phenyl]methanamine

  • MDL number MFCD19612527
  • Cas Number : 1341447-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189965
Availability details for
Quantity
  • Availability
  • Add to Cart