ENA429360507 Enamine

5-(2-methylphenyl)imidazolidine-2,4-dione

  • MDL number MFCD09729961
  • Cas Number : 187978-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189967
Availability details for
Quantity
  • Availability
  • Add to Cart