ENA429360510 Enamine

1-(piperidin-2-yl)propan-2-ol

  • MDL number MFCD13184003
  • Cas Number : 10047-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189971
Availability details for
Quantity
  • Availability
  • Add to Cart