ENA429360512 Enamine

4-chloro-N1,N1-dimethylbenzene-1,3-disulfonamide

  • MDL number MFCD11212965
  • Cas Number : 14748-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189973
Availability details for
Quantity
  • Availability
  • Add to Cart