ENA429360513 Enamine

6-bromo-7-sulfanyl-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD11206336
  • Cas Number : 1094268-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189974
Availability details for
Quantity
  • Availability
  • Add to Cart