ENA429360515 Enamine

1-(3-aminophenyl)-1,3-dimethylurea

  • MDL number MFCD24262214
  • Cas Number : 1545161-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189977

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189977
Availability details for
Quantity
  • Availability
  • Add to Cart