ENA429360518 Enamine

3-bromo-4-cyanobenzene-1-sulfonamide

  • MDL number MFCD18390723
  • Cas Number : 1261726-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189489

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189489
Availability details for
Quantity
  • Availability
  • Add to Cart