ENA429360519 Enamine

2-chloro-4-sulfamoylbenzamide

  • MDL number MFCD11650754
  • Cas Number : 1099660-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189987
Availability details for
Quantity
  • Availability
  • Add to Cart