ENA429360523 Enamine

2-cyano-3-(4-cyanophenyl)propanoic acid

  • MDL number MFCD24487782
  • Cas Number : 18176-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190034
Availability details for
Quantity
  • Availability
  • Add to Cart