ENA429360524 Enamine

5-fluoro-2-(thiomorpholin-4-yl)benzonitrile

  • MDL number MFCD11041626
  • Cas Number : 1272094-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190038
Availability details for
Quantity
  • Availability
  • Add to Cart