ENA429360526 Enamine

2-(2,2-difluoroethyl)-2,3-dihydro-1H-isoindol-5-amine

  • MDL number MFCD21664799
  • Cas Number : 1484193-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190063

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190063
Availability details for
Quantity
  • Availability
  • Add to Cart