ENA429360527 Enamine

4-ethyl-3-(trifluoromethyl)-1H-pyrazol-5-amine

  • MDL number MFCD11558351

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190072

Safety Information

msds https://www.enaminestore.com/catalog/EN300-190072
Availability details for
Quantity
  • Availability
  • Add to Cart