ENA429360528 Enamine

5-(oxan-3-yl)-1H-pyrazol-3-amine

  • MDL number MFCD28390172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-190073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-190073
Availability details for
Quantity
  • Availability
  • Add to Cart